siRNA Details
Gene Symbol: Brca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031542.2 | NP_113730.2 | 3 | 269 | ||
GenBank mRNA | U89653.1 | - | 3 | 268 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Brca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031542.2 | NP_113730.2 | 3 | 269 | ||
GenBank mRNA | U89653.1 | - | 3 | 268 |