siRNA Details
| Gene Symbol: IFIT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012420.2 | NP_036552.1 | 2 | 3323 | ||
| GenBank mRNA | U34605.1 | - | 2 | 3199 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IFIT5| Gene Symbol: IFIT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012420.2 | NP_036552.1 | 2 | 3323 | ||
| GenBank mRNA | U34605.1 | - | 2 | 3199 | ||