siRNA Details
| Gene Symbol: Ampd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031544.1 | NP_113732.1 | 15 | 2896 | ||
| GenBank mRNA | U90888.1 | - | 15 | 2896 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ampd3| Gene Symbol: Ampd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031544.1 | NP_113732.1 | 15 | 2896 | ||
| GenBank mRNA | U90888.1 | - | 15 | 2896 | ||