siRNA Details
| Gene Symbol: Scnn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031548.2 | NP_113736.1 | 12 | 3368 | ||
| GenBank mRNA | BC070902.1 | - | 12 | 3368 | 7100777 | |
| X70497.1 | - | 12 | 3079 | |||
| X70521.1 | - | 12 | 3056 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scnn1a| Gene Symbol: Scnn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031548.2 | NP_113736.1 | 12 | 3368 | ||
| GenBank mRNA | BC070902.1 | - | 12 | 3368 | 7100777 | |
| X70497.1 | - | 12 | 3079 | |||
| X70521.1 | - | 12 | 3056 | |||