siRNA Details
| Gene Symbol: Cyp2c12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031572.1 | NP_113760.1 | 5 | 690 | ||
| GenBank mRNA | FQ209627.1 | - | 2 | 129 | ||
| FQ219038.1 | - | 3 | 371 | |||
| FQ219491.1 | - | 4 | 565 | |||
| FQ219720.1 | - | 2 | 160 | |||
| J03786.1 | - | 5 | 690 | |||

Cyp2c12
Genomic Map