siRNA Details
| Gene Symbol: Calm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031969.2 | NP_114175.1 | 6 | 903 | ||
| GenBank mRNA | AF178845.1 | - | 838 | |||
| FQ213013.1 | - | 2 | 466 | |||
| FQ213227.1 | - | 6 | 1018 | |||
| FQ217595.1 | - | 1 | 42 | |||
| FQ225380.1 | - | 6 | 932 | |||
| FQ226971.1 | - | 6 | 977 | |||
| FQ227186.1 | - | 6 | 974 | |||
| FQ232999.1 | - | 3 | 536 | |||
| X13933.1 | - | 6 | 903 | |||

Calm1
Genomic Map