siRNA Details
| Gene Symbol: Hagh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033349.2 | NP_203500.2 | 5 | 526 | ||
| XM_006245899.1 | XP_006245961.1 | 5 | 503 | |||
| XM_006245900.1 | XP_006245962.1 | 4 | 417 | |||
| GenBank mRNA | BC097301.1 | - | 5 | 478 | ||
| FQ229806.1 | - | 5 | 595 | |||

Hagh
Genomic Map