siRNA Details
| Gene Symbol: Gata2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033442.1 | NP_254277.1 | 4 | 1110 | ||
| GenBank mRNA | AB102789.1 | - | 2 | 667 | ||
| AB102790.1 | - | 2 | 667 | |||
| AF345897.1 | - | 2 | 667 | |||
| AY032734.1 | - | 4 | 1110 | |||
| BC061745.1 | - | 3 | 917 | 5599740 | ||
| FQ212304.1 | - | 991 | ||||

Gata2
Genomic Map