siRNA Details
| Gene Symbol: Gata2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033442.1 | NP_254277.1 | 7 | 2739 | ||
| GenBank mRNA | AY032734.1 | - | 7 | 2739 | ||
| BC061745.1 | - | 6 | 2546 | 5599740 | ||
| FQ212304.1 | - | 2619 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gata2| Gene Symbol: Gata2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033442.1 | NP_254277.1 | 7 | 2739 | ||
| GenBank mRNA | AY032734.1 | - | 7 | 2739 | ||
| BC061745.1 | - | 6 | 2546 | 5599740 | ||
| FQ212304.1 | - | 2619 | ||||