siRNA Details
| Gene Symbol: Kap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052802.1 | NP_434689.1 | 4 | 473 | ||
| XM_006237483.2 | XP_006237545.1 | 3 | 338 | |||
| GenBank mRNA | U25808.1 | - | 4 | 473 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kap| Gene Symbol: Kap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052802.1 | NP_434689.1 | 4 | 473 | ||
| XM_006237483.2 | XP_006237545.1 | 3 | 338 | |||
| GenBank mRNA | U25808.1 | - | 4 | 473 | ||