siRNA Details
| Gene Symbol: Atp7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052803.2 | NP_434690.1 | 3 | 660 | ||
| XM_006256995.2 | XP_006257057.1 | 4 | 827 | |||
| XM_006256997.2 | XP_006257059.1 | 4 | 827 | |||
| XM_008773334.1 | XP_008771556.1 | 4 | 768 | |||
| GenBank mRNA | U59245.1 | - | 3 | 665 | ||

Atp7a
Genomic Map