siRNA Details
| Gene Symbol: Fmr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052804.1 | NP_434691.1 | 4 | 414 | ||
| XM_006229530.2 | XP_006229592.1 | 4 | 464 | |||
| XM_006229531.2 | XP_006229593.1 | 4 | 464 | |||
| XM_006229532.2 | XP_006229594.1 | 4 | 464 | |||
| XM_006229535.2 | XP_006229597.1 | 3 | 183 | |||
| XM_008773597.1 | XP_008771819.1 | 4 | 296 | |||
| GenBank mRNA | AF435434.1 | - | 4 | 233 | ||
| AY240947.1 | - | 4 | 414 | |||
| U60145.1 | - | 2 | 123 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008031, AK053701, AK080948, AK053211, AK140343, and others.

Fmr1
Genomic Map