siRNA Details
| Gene Symbol: Chrna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | L31621.1 | - | 6 | 1688 | ||
| X03440.1 | - | 6 | 1762 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrna3| Gene Symbol: Chrna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | L31621.1 | - | 6 | 1688 | ||
| X03440.1 | - | 6 | 1762 | |||