siRNA Details
| Gene Symbol: Avpr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053019.2 | NP_444178.2 | 1 | 510 | ||
| GenBank mRNA | BC088095.1 | - | 1 | 494 | 7321427 | |
| Z11690.1 | - | 1 | 518 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Avpr1a| Gene Symbol: Avpr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053019.2 | NP_444178.2 | 1 | 510 | ||
| GenBank mRNA | BC088095.1 | - | 1 | 494 | 7321427 | |
| Z11690.1 | - | 1 | 518 | |||
Set Membership: |
|