siRNA Details
| Gene Symbol: Reg3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053289.1 | NP_445741.1 | 6 | 590 | ||
| GenBank mRNA | FQ232415.1 | - | 6 | 590 | ||
| FQ233015.1 | - | 6 | 590 | |||
| M55149.1 | - | 6 | 592 | |||
| M98049.1 | - | 5 | 540 | |||
| S43715.1 | - | 6 | 590 | |||

Reg3b
Genomic Map