siRNA Details
| Gene Symbol: SSBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256732.1 | NP_001243661.1 | 3 | 394 | ||
| NM_001256733.1 | NP_001243662.1 | 3 | 394 | |||
| NM_001256734.1 | NP_001243663.1 | 3 | 394 | |||
| NM_001256735.1 | NP_001243664.1 | 3 | 394 | |||
| NM_001256736.1 | NP_001243665.1 | 3 | 394 | |||
| NM_012446.3 | NP_036578.2 | 3 | 394 | |||
| XM_011543297.2 | XP_011541599.1 | 3 | 309 | |||
| XM_017009304.1 | XP_016864793.1 | 3 | 212 | |||
| XM_017009305.1 | XP_016864794.1 | 4 | 325 | |||
| XM_017009306.1 | XP_016864795.1 | 3 | 309 | |||
| XM_017009307.1 | XP_016864796.1 | 3 | 212 | |||
| XM_017009308.1 | XP_016864797.1 | 4 | 473 | |||
| XM_017009309.1 | XP_016864798.1 | 3 | 537 | |||
| XM_017009310.1 | XP_016864799.1 | 3 | 685 | |||
| GenBank mRNA | AF077048.1 | - | 3 | 231 | ||
| AF161465.1 | - | 3 | 224 | |||
| AK074537.1 | - | 3 | 189 | |||
| AK293544.1 | - | 3 | 189 | |||
| AK294864.1 | - | 3 | 189 | |||
| AK297494.1 | - | 3 | 189 | |||
| AK299858.1 | - | 3 | 238 | |||
| AK312337.1 | - | 3 | 238 | |||
| AL080076.1 | - | 3 | 242 | |||
| BC017020.1 | - | 3 | 242 | 3913337 | ||
| BI517313.1 | - | 3 | 394 | |||
| CR936787.1 | - | 3 | 239 | |||

SSBP2
Genomic Map