siRNA Details
| Gene Symbol: Aurkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053749.1 | NP_446201.1 | 5 | 411 | ||
| XM_006246569.2 | XP_006246631.1 | 5 | 429 | |||
| XM_006246570.2 | XP_006246632.1 | 4 | 316 | |||
| XM_008767757.1 | XP_008765979.1 | 5 | 326 | |||
| XM_008767758.1 | XP_008765980.1 | 5 | 272 | |||
| GenBank mRNA | BC097297.1 | - | 5 | 329 | 7454285 | |
| D89731.1 | - | 5 | 411 | |||

Aurkb
Genomic Map