siRNA Details
| Gene Symbol: Smu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057195.1 | NP_476543.1 | 2 | 134 | ||
| GenBank mRNA | AY029526.1 | - | 2 | 134 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smu1| Gene Symbol: Smu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057195.1 | NP_476543.1 | 2 | 134 | ||
| GenBank mRNA | AY029526.1 | - | 2 | 134 | ||