siRNA Details
| Gene Symbol: Ada | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130399.2 | NP_569083.1 | 6 | 644 | ||
| GenBank mRNA | AB059655.1 | - | 6 | 568 | ||
| BC088116.1 | - | 6 | 644 | 7384286 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ada| Gene Symbol: Ada | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130399.2 | NP_569083.1 | 6 | 644 | ||
| GenBank mRNA | AB059655.1 | - | 6 | 568 | ||
| BC088116.1 | - | 6 | 644 | 7384286 | ||