siRNA Details
| Gene Symbol: Zdhhc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133394.1 | NP_596885.1 | 3 | 603 | ||
| GenBank mRNA | AY040615.1 | - | 3 | 603 | ||
| AY886525.1 | - | 3 | 598 | |||
| BC061769.1 | - | 2 | 379 | 5621307 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zdhhc7| Gene Symbol: Zdhhc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133394.1 | NP_596885.1 | 3 | 603 | ||
| GenBank mRNA | AY040615.1 | - | 3 | 603 | ||
| AY886525.1 | - | 3 | 598 | |||
| BC061769.1 | - | 2 | 379 | 5621307 | ||