siRNA Details
Gene Symbol: Zfp483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133422.1 | NP_596913.1 | 6 | 1329 | ||
GenBank mRNA | AF277901.1 | - | 6 | 1329 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133422.1 | NP_596913.1 | 6 | 1329 | ||
GenBank mRNA | AF277901.1 | - | 6 | 1329 |