siRNA Details
| Gene Symbol: Cyb5r4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133427.3 | NP_596918.3 | 3, 4 | 461 | ||
| XM_006243472.2 | XP_006243534.1 | 3, 4 | 477 | |||
| XM_008766437.1 | XP_008764659.1 | 3, 4 | 620 | |||
| GenBank mRNA | AF307840.1 | - | 2, 3 | 220 | ||
| AY321370.1 | - | 2, 3 | 220 | |||
| AY321371.1 | - | 2, 3 | 220 | |||
| BC080240.1 | - | 3, 4 | 439 | 7099950 | ||

Cyb5r4
Genomic Map