siRNA Details
| Gene Symbol: Adcyap1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270579.1 | NP_001257508.1 | 18 | 2108 | ||
| NM_001270580.1 | NP_001257509.1 | 17 | 2024 | |||
| NM_001270581.1 | NP_001257510.1 | 17 | 2052 | |||
| NM_001270582.1 | NP_001257511.1 | 16 | 1940 | |||
| NM_001270583.1 | NP_001257512.1 | 17 | 2021 | |||
| NM_133511.2 | NP_598195.1 | 17 | 2024 | |||
| XM_008762870.1 | XP_008761092.1 | 16 | 1767 | |||
| GenBank mRNA | D14908.1 | - | 17 | 1932 | ||
| D14909.1 | - | 17 | 1814 | |||
| D16465.1 | - | 17 | 2024 | |||
| L16506.1 | - | 11 | 1344 | |||
| L16680.1 | - | 16 | 1716 | |||
| Z22735.1 | - | 11 | 1344 | |||
| Z23282.1 | - | 17 | 2080 | |||
More Information
Set Membership: |
|

Adcyap1r1
Genomic Map