siRNA Details
| Gene Symbol: Klrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133512.2 | NP_598196.1 | 8 | 1143 | ||
| XM_006237073.2 | XP_006237135.1 | 8 | 2325 | |||
| XM_006237074.2 | XP_006237136.1 | 8 | 1224 | |||
| XM_006237075.2 | XP_006237137.1 | 8 | 1293 | |||
| XM_006237076.2 | XP_006237138.1 | 8 | 1131 | |||
| XM_006237077.2 | XP_006237139.1 | 8 | 1181 | |||
| GenBank mRNA | AF009511.1 | - | 7 | 1097 | ||

Klrk1
Genomic Map