siRNA Details
| Gene Symbol: Rdh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133543.3 | NP_598227.3 | 746 | |||
| GenBank mRNA | BC088090.1 | - | 746 | 7368214 | ||
| BC098650.1 | - | 808 | 7371059 | |||
| FQ209576.1 | - | 730 | ||||
| U18762.1 | - | 955 | ||||
| U33501.1 | - | 727 | ||||

Rdh7
Genomic Map