siRNA Details
| Gene Symbol: Pla2g4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133551.2 | NP_598235.2 | 5, 6 | 554 | ||
| GenBank mRNA | BC070940.1 | - | 5, 6 | 554 | 7099813 | |
| S77829.1 | - | 5, 6 | 540 | |||
| U38376.1 | - | 5, 6 | 555 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g4a| Gene Symbol: Pla2g4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133551.2 | NP_598235.2 | 5, 6 | 554 | ||
| GenBank mRNA | BC070940.1 | - | 5, 6 | 554 | 7099813 | |
| S77829.1 | - | 5, 6 | 540 | |||
| U38376.1 | - | 5, 6 | 555 | |||