siRNA Details
| Gene Symbol: Atg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134394.2 | NP_599221.1 | 4, 5 | 517 | ||
| GenBank mRNA | AF175224.1 | - | 4, 5 | 520 | ||
| BC078743.1 | - | 4, 5 | 529 | 7112937 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atg3| Gene Symbol: Atg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134394.2 | NP_599221.1 | 4, 5 | 517 | ||
| GenBank mRNA | AF175224.1 | - | 4, 5 | 520 | ||
| BC078743.1 | - | 4, 5 | 529 | 7112937 | ||