siRNA Details
| Gene Symbol: Agt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134432.2 | NP_602308.1 | 2 | 841 | ||
| XM_008772597.1 | XP_008770819.1 | 3 | 1041 | |||
| GenBank mRNA | BC078741.1 | - | 2 | 841 | 7128542 | |
| BC087679.1 | - | 1 | 677 | 7318795 | ||

Agt
Genomic Map