siRNA Details
| Gene Symbol: Dab2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138710.3 | NP_619724.3 | 6 | 826 | ||
| XM_006234057.2 | XP_006234119.1 | 5 | 813 | |||
| XM_008761697.1 | XP_008759919.1 | 3 | 1224 | |||
| XM_008761698.1 | XP_008759920.1 | 3 | 1224 | |||
| XM_008761699.1 | XP_008759921.1 | 3 | 1225 | |||
| GenBank mRNA | AF236130.1 | - | 5 | 534 | ||
| BC061865.1 | - | 2 | 140 | 5598046 | ||

Dab2ip
Genomic Map