siRNA Details
Gene Symbol: Serpina12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138825.2 | NP_620180.2 | 2 | 774 | ||
GenBank mRNA | AF245398.1 | - | 2 | 774 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Serpina12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138825.2 | NP_620180.2 | 2 | 774 | ||
GenBank mRNA | AF245398.1 | - | 2 | 774 |