siRNA Details
Gene Symbol: Csn1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138874.2 | NP_620229.2 | 5 | 186 | ||
GenBank mRNA | J00710.1 | - | 5 | 186 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Csn1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138874.2 | NP_620229.2 | 5 | 186 | ||
GenBank mRNA | J00710.1 | - | 5 | 186 |