siRNA Details
| Gene Symbol: Ampd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138876.1 | NP_620231.1 | 3 | 208 | ||
| XM_006233060.2 | XP_006233122.1 | 2 | 285 | |||
| XM_008761369.1 | XP_008759591.1 | 2 | 208 | |||
| GenBank mRNA | J02811.1 | - | 3 | 208 | ||

Ampd1
Genomic Map