siRNA Details
| Gene Symbol: TSPAN16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282509.1 | NP_001269438.1 | 2, 3 | 420 | ||
| NM_012466.3 | NP_036598.1 | 2, 3 | 420 | |||
| NR_104199.1 | 2, 3 | 420 | ||||
| XM_011527901.1 | XP_011526203.1 | 2, 3 | 415 | |||
| XM_011527902.1 | XP_011526204.1 | 2, 3 | 415 | |||
| XM_011527903.2 | XP_011526205.1 | 2, 3 | 351 | |||
| GenBank mRNA | BC029908.1 | - | 2, 3 | 390 | 5168677 | |
| BM560276.1 | - | 2, 3 | 407 | |||
| DB457828.1 | - | 2, 3 | 410 | |||

TSPAN16
Genomic Map