siRNA Details
Gene Symbol: Cgref1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139087.2 | NP_620787.2 | 6 | 980 | ||
GenBank mRNA | FQ220335.1 | - | 6 | 910 | ||
U66470.1 | - | 6 | 901 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cgref1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139087.2 | NP_620787.2 | 6 | 980 | ||
GenBank mRNA | FQ220335.1 | - | 6 | 910 | ||
U66470.1 | - | 6 | 901 |