siRNA Details
| Gene Symbol: Impg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139088.2 | NP_620788.2 | 8 | 850 | ||
| XM_008768628.1 | XP_008766850.1 | 8 | 1838 | |||
| XM_008768629.1 | XP_008766851.1 | 3 | 198 | |||
| GenBank mRNA | U76717.1 | - | 8 | 850 | ||

Impg2
Genomic Map