siRNA Details
| Gene Symbol: TPSG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012467.3 | NP_036599.3 | 6 | 982 | ||
| XM_011522446.2 | XP_011520748.1 | 6 | 1721 | |||
| XM_011522447.2 | XP_011520749.1 | 6 | 2595 | |||
| GenBank mRNA | BE552396.1 | - | 1 | 244 | ||

TPSG1
Genomic Map