siRNA Details
| Gene Symbol: Arfgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145090.3 | NP_659558.1 | 4 | 398 | ||
| XM_006235731.2 | XP_006235793.1 | 4 | 498 | |||
| XM_006235732.1 | XP_006235794.1 | 4 | 439 | |||
| XM_006235733.2 | XP_006235795.1 | 3 | 707 | |||
| XM_006235734.2 | XP_006235796.1 | 3 | 706 | |||
| GenBank mRNA | BC070895.1 | - | 4 | 379 | 7099840 | |
| DQ167810.1 | - | 2 | 115 | |||
| DQ167811.1 | - | 2 | 115 | |||
| FQ222295.1 | - | 4 | 421 | |||
| FQ232032.1 | - | 4 | 419 | |||
| FQ233265.1 | - | 4 | 419 | |||
| U35776.1 | - | 4 | 398 | |||

Arfgap1
Genomic Map