siRNA Details
| Gene Symbol: Carhsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152790.2 | NP_690003.1 | 4 | 420 | ||
| XM_006245750.2 | XP_006245812.1 | 4 | 1203 | |||
| XM_006245753.2 | XP_006245815.1 | 4 | 574 | |||
| XM_008767479.1 | XP_008765701.1 | 4 | 897 | |||
| GenBank mRNA | AF115346.1 | - | 3 | 390 | ||
| BC071173.1 | - | 4 | 420 | 7099252 | ||

Carhsp1
Genomic Map