siRNA Details
Gene Symbol: Gpr156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153295.1 | NP_695207.1 | 1, 2 | 100 | ||
GenBank mRNA | AF488741.1 | - | 1, 2 | 100 |
Gene Symbol: LOC103693583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768769.1 | XP_008766991.1 | 2, 3 | 794 |
