siRNA Details
| Gene Symbol: Gpr156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153295.1 | NP_695207.1 | 3, 4 | 360 | ||
| GenBank mRNA | AF488741.1 | - | 3, 4 | 360 | ||
| Gene Symbol: LOC103693583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768769.1 | XP_008766991.1 | 4, 5 | 1054 | ||

Gpr156
Genomic Map