siRNA Details
| Gene Symbol: Gpr156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153295.1 | NP_695207.1 | 8 | 974 | ||
| GenBank mRNA | AF488741.1 | - | 8 | 974 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr156| Gene Symbol: Gpr156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153295.1 | NP_695207.1 | 8 | 974 | ||
| GenBank mRNA | AF488741.1 | - | 8 | 974 | ||