siRNA Details
| Gene Symbol: Mina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153309.2 | NP_695221.1 | 2 | 331 | ||
| XM_008768680.1 | XP_008766902.1 | 2 | 396 | |||
| GenBank mRNA | AB083195.1 | - | 1 | 306 | ||
| BC087650.1 | - | 2 | 331 | 7310399 | ||
| Gene Symbol: LOC689439 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761010.1 | XP_008759232.1 | 1 | 374 | ||

Mina
Genomic Map