siRNA Details
Gene Symbol: Slc13a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170668.1 | NP_733768.1 | 2, 3 | 259 | ||
XM_006246602.2 | XP_006246664.1 | 3, 4 | 1485 | |||
GenBank mRNA | AF522186.1 | - | 2, 3 | 259 |
