siRNA Details
| Gene Symbol: Slc13a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170668.1 | NP_733768.1 | 2, 3 | 262 | ||
| XM_006246602.2 | XP_006246664.1 | 3, 4 | 1488 | |||
| GenBank mRNA | AF522186.1 | - | 2, 3 | 262 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc13a5| Gene Symbol: Slc13a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170668.1 | NP_733768.1 | 2, 3 | 262 | ||
| XM_006246602.2 | XP_006246664.1 | 3, 4 | 1488 | |||
| GenBank mRNA | AF522186.1 | - | 2, 3 | 262 | ||