siRNA Details
| Gene Symbol: Arid1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758745.1 | XP_008756967.1 | 1 | 1849 | ||
| GenBank mRNA | AJ005202.2 | - | 4416 | |||
| AJ440711.1 | - | 5581 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arid1b| Gene Symbol: Arid1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758745.1 | XP_008756967.1 | 1 | 1849 | ||
| GenBank mRNA | AJ005202.2 | - | 4416 | |||
| AJ440711.1 | - | 5581 | ||||