siRNA Details
| Gene Symbol: Vom1r101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173112.2 | NP_775135.2 | 1 | 556 | ||
| GenBank mRNA | AY510287.1 | - | 1 | 556 | ||
| U36786.1 | - | 1 | 474 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r101| Gene Symbol: Vom1r101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173112.2 | NP_775135.2 | 1 | 556 | ||
| GenBank mRNA | AY510287.1 | - | 1 | 556 | ||
| U36786.1 | - | 1 | 474 | |||