siRNA Details
| Gene Symbol: Havcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173149.2 | NP_775172.1 | 2 | 959 | ||
| XM_006246163.2 | XP_006246225.1 | 3 | 604 | |||
| XM_008767665.1 | XP_008765887.1 | 4 | 656 | |||
| XM_008767666.1 | XP_008765888.1 | 3 | 1241 | |||
| GenBank mRNA | AF035963.1 | - | 2 | 960 | ||
| BC061820.1 | - | 3 | 399 | 5599247 | ||

Havcr1
Genomic Map