siRNA Details
| Gene Symbol: Pcyt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173151.1 | NP_775174.1 | 4 | 366 | ||
| XM_006257016.2 | XP_006257078.1 | 4 | 327 | |||
| XM_006257017.2 | XP_006257079.1 | 4 | 734 | |||
| XM_008773254.1 | XP_008771476.1 | 4 | 447 | |||
| GenBank mRNA | AF190256.1 | - | 4 | 366 | ||

Pcyt1b
Genomic Map