siRNA Details
| Gene Symbol: Yy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173290.1 | NP_775412.1 | 2 | 821 | ||
| XM_006240537.2 | XP_006240599.1 | 2 | 1621 | |||
| GenBank mRNA | AB080317.1 | - | 2 | 821 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Yy1| Gene Symbol: Yy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173290.1 | NP_775412.1 | 2 | 821 | ||
| XM_006240537.2 | XP_006240599.1 | 2 | 1621 | |||
| GenBank mRNA | AB080317.1 | - | 2 | 821 | ||