siRNA Details
| Gene Symbol: Olr1271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173300.1 | NP_775422.1 | 2 | 1133 | ||
| GenBank mRNA | X80671.1 | - | 2 | 1133 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1271| Gene Symbol: Olr1271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173300.1 | NP_775422.1 | 2 | 1133 | ||
| GenBank mRNA | X80671.1 | - | 2 | 1133 | ||